Skip to Content
Merck
All Photos(1)

Key Documents

L511838

Sigma-Aldrich

[Ir(dtbbpy)(ppy)2][PF6]

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H40F6IrN4P
CAS Number:
Molecular Weight:
913.95
UNSPSC Code:
12352200
PubChem Substance ID:

reaction suitability

reagent type: catalyst
reaction type: C-H Activation

SMILES string

CC(C1=CC=[N]([Ir]2([N]3=CC=C(C=C34)C(C)(C)C)([N]5=CC=CC=C5C6=CC=CC=C62)(C7=CC=CC=C78)[N]9=C8C=CC=C9)C4=C1)(C)C.FP(F)(F)(F)(F)F

InChI

1S/C18H24N2.2C11H8N.F6P.Ir/c1-17(2,3)13-7-9-19-15(11-13)16-12-14(8-10-20-16)18(4,5)6;2*1-2-6-10(7-3-1)11-8-4-5-9-12-11;1-7(2,3,4,5)6;/h7-12H,1-6H3;2*1-6,8-9H;;

InChI key

DZARMRZNOUWGQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for the visible-light mediated formation of alpha-amino radicals, used by Nishibayashi for addition into electron-poor alkenes and imines

Visible-Light-Mediated Utilization of ·-Aminoalkyl Radicals: Addition to Electron-Deficient Alkenes Using Photoredox Catalysts

In presence of [Co(bpy)3]2+, serves as photosensitizer for H2 generation from water (water splitting).

Tricyclometalated Iridium Complexes as Highly Stable Photosensitizers for Light-Induced Hydrogen Evolution

For other Visible-Light Photocatalysts, refer to: www.aldrich.com/photocatalysis

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service