J100006
Bis(N,N′-di-tert-butylacetamidinato)iron(II)
electronic grade, ≥99.99% metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H42N4Fe
CAS Number:
Molecular Weight:
394.42
UNSPSC Code:
12352300
grade
electronic grade
vapor pressure
0.06 mmHg ( 55 °C)
Assay
≥99.99% metals basis
form
solid
color
white
bp
60 °C/0.02 mmHg
mp
105-109 °C (lit.)
107 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service