GF62341321
Monel® alloy 400, Ni65Cu33Fe2
500mm tube, 2.8mm outside diameter, 0.35mm wall thickness, 2.1mm inside diameter, as drawn
Synonym(s):
NI117450
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352300
NACRES:
NA.23
form
tubes
manufacturer/tradename
Goodfellow 623-413-21
L × wall thickness
500 mm × 0.35 mm
O.D. × I.D.
2.8 mm × 2.1 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Monel is a registered trademark of Special Metals Corporation
Monel is a registered trademark of Huntington Alloys Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service