Skip to Content
Merck
All Photos(2)

Documents

GF56438714

Nickel

tube, 1000mm, outside diameter 2.5mm, inside diameter 2.25mm, wall thickness 0.125mm, hard, 99.5%

Synonym(s):

Nickel, NI007340

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5%

form

tubes

manufacturer/tradename

Goodfellow 564-387-14

resistivity

6.97 μΩ-cm, 20°C

L × wall thickness

1000 mm × 0.125 mm

O.D. × I.D.

2.5 mm × 2.25 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erkai He et al.
Environmental pollution (Barking, Essex : 1987), 188, 17-26 (2014-02-18)
Protons and other cations may inhibit metal uptake and alleviate metal toxicity in aquatic organisms, but less is known about these interactions in soil organisms. The present study investigated the influence of solution chemistry on uptake and toxicity of Ni
Gary A Molander et al.
Organic letters, 16(7), 1904-1907 (2014-03-29)
A general method for the alkenylation of alkyl electrophiles using nearly stoichiometric amounts of the air- and moisture-stable potassium organotrifluoroborates has been developed. Various functional groups were tolerated on both the nucleophilic and electrophilic partner. Reactions of highly substituted E-
Filaggrin is a predominant member of the denaturation-resistant nickel-binding proteome of human epidermis.
Katrine Ross-Hansen et al.
The Journal of investigative dermatology, 134(4), 1164-1166 (2013-10-26)
Meinar Nur Ashrin et al.
Journal of immunology (Baltimore, Md. : 1950), 192(9), 4025-4031 (2014-03-29)
Ni is the most frequent cause of contact allergy induced by metals. However, the underlying mechanism of this induction is unknown. Our previous research demonstrates that activation of dendritic cells (DCs) through p38MAPK/MKK6 is required for Ni-induced allergy in mice.
Sébastien Hulo et al.
Toxicology letters, 226(1), 63-69 (2014-02-11)
To evaluate manganese in exhaled breath condensate (Mn-EBC) as an indicator of exposure to fumes from metal inert gas welding process. We collected EBC and urine from 17 welders and 16 unexposed control subjects after 5 days exposure. Concentrations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service