GF41844067
Havar® - high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn
foil, 150 x 150mm, 0.075mm thickness, as rolled, light tight (lt)
Synonym(s):
CO010300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
light tight (lt)
form
foil (as rolled)
manufacturer/tradename
Goodfellow 418-440-67
size × thickness
150 mm × 150 mm × 0.075 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Havar is a registered trademark of Precision Metals, Inc. AMETEK, or its subsidiaries or affiliates.
Havar is a registered trademark of Precision Metals, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service