GF11738842
Zamak 410 - zinc/aluminum/copper/magnesium
foam, Zn 95%/Al 4%/Cu 1%/Mg, thickness 20 mm, size 150 × 150 mm, bulk density 0.33 g/cm3
Synonym(s):
ZN063801
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352311
NACRES:
NA.23
Recommended Products
Assay
≥95%
manufacturer/tradename
Goodfellow 117-388-42
size
150 × 150 mm
thickness
20 mm
bulk density
0.33 g/cm3
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service