Skip to Content
Merck
All Photos(2)

Key Documents

GF01198762

Cadmium

wire reel, 1m, diameter 0.25mm, hard, 99.99+%

Synonym(s):

Cadmium, CD005100

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
2.520,00 kr.

2.520,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
2.520,00 kr.

About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein:
8137359
MDL number:
UNSPSC Code:
12141707
PubChem Substance ID:
NACRES:
NA.23

2.520,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 011-987-62

resistivity

7.27 μΩ-cm, 22°C

L × diam.

1 m × 0.25 mm

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peerapat Kosolsaksakul et al.
Environmental pollution (Barking, Essex : 1987), 187, 153-161 (2014-02-08)
The Mae Tao watershed, northwest Thailand, has become contaminated with cadmium (Cd) as a result of zinc ore extraction (Padaeng deposit) in the nearby Thanon-Thongchai mountains. Consumption of contaminated rice has led to documented human health impacts. The aim of
Jian-An Xian et al.
Bulletin of environmental contamination and toxicology, 92(5), 525-528 (2014-01-01)
This study investigated the effect of ambient Cadmium (Cd) on haemocyte apoptosis of the shrimp, Penaeus monodon. Cellular response was determined in Cd-exposed (0, 0.05, 0.5 and 5 mg L(-1)) shrimp. Results showed that 0.05 mg L(-1) Cd(2+) had no significant effect on the
Lijun Li et al.
Environmental pollution (Barking, Essex : 1987), 187, 90-97 (2014-01-28)
We studied the bonding and release kinetics of Cd, Cu and Pb from different soils in the older metropolitan area of Copenhagen. Total Cd, Cu and Pb concentrations were elevated 5-27 times in the urban soils compared to an agricultural
Jachym Suman et al.
Biochimica et biophysica acta, 1838(5), 1338-1343 (2014-02-05)
PbtA, a putative P(1B)-type ATPase from the Gram-negative soil bacterium Achromobacter xylosoxidans A8 responsible for Pb(2+)/Zn(2+)/Cd(2+)-resistance in Escherichia coli, was heterologously expressed in Saccharomyces cerevisiae. When present in Zn(2+)- and Pb(2+)/Cd(2+)-hypersensitive S. cerevisiae strains CM137 and DTY168, respectively, PbtA was
S E Dal-Medico et al.
Ecotoxicology and environmental safety, 104, 168-174 (2014-04-01)
A drastic amphibian decline has been observed worldwide, which can be attributed (among other factors) to exposure to pollutants. Considering that cadmium corresponds to the most rapidly increasing trace metal in the environment, the aim of this work was to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service