E47255
Ethyl 3-pyridylacetate
99%
Synonym(s):
Ethyl 3-pyridineacetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Beilstein:
123827
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.500 (lit.)
bp
80 °C/0.4 mmHg (lit.)
density
1.086 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)Cc1cccnc1
InChI
1S/C9H11NO2/c1-2-12-9(11)6-8-4-3-5-10-7-8/h3-5,7H,2,6H2,1H3
InChI key
RPWXYCRIAGBAGY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service