Skip to Content
Merck
All Photos(1)

Key Documents

E35702

Sigma-Aldrich

Ethyl 4-methyl-2-cyclohexanone-1-carboxylate

Synonym(s):

4-Methyl-2-oxocyclohexanecarboxylic acid ethyl ester, Ethyl 4-methyl-2-oxo-1-cyclohexanecarboxylate, NSC 84213

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H8(=O)CO2C2H5
CAS Number:
Molecular Weight:
184.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

impurities

10% methyl ester

refractive index

n20/D 1.473 (lit.)

bp

128-132 °C/32 mmHg (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1CCC(C)CC1=O

InChI

1S/C10H16O3/c1-3-13-10(12)8-5-4-7(2)6-9(8)11/h7-8H,3-6H2,1-2H3

InChI key

FFVHHWOJYFCNJP-UHFFFAOYSA-N

Application

Reactant involved in:
  • Hydroxylation / oxidative chlorination with hydrogen peroxide
  • Heterocyclization
  • Cyclization of generated dianions followed by dehydrogenation
  • Palladium-catalyzed isomerization
  • Regioselective cyclizations
  • Vinylogous Michael reactions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service