All Photos(1)
About This Item
Linear Formula:
CH3COCH(CH2C6H5)CO2C2H5
CAS Number:
Molecular Weight:
220.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.5 (lit.)
bp
276 °C (lit.)
density
1.036 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)C(Cc1ccccc1)C(C)=O
InChI
1S/C13H16O3/c1-3-16-13(15)12(10(2)14)9-11-7-5-4-6-8-11/h4-8,12H,3,9H2,1-2H3
InChI key
XDWQYMXQMNUWID-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service