Skip to Content
Merck
All Photos(1)

Documents

DWKFDF030N

Azlon Polypropylene Volumetric Flask With PPE Stopper

class B, capacity 250 mL

Synonym(s):

FDF030N

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

feature

class B

packaging

pkg of 1 ea

manufacturer/tradename

Azlon FDF030N

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

General description

With an easy to read calibration line, the AZLON® class B volumetric flask is suitable for accurate measuring and is supplied with a polypropylene stopper. Provide a good alternative to glass.

Application

For the accurate preparation of solutions where glass alternatives would not be suitable

Features and Benefits

Excellent durability and calibrated to ISO 1042

Legal Information

AZLON is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service