Skip to Content
Merck
All Photos(1)

Key Documents

DWKBGS042

Azlon Polyamide Coated Steel Bottle Carrier

to hold, up to six 74mm diameter bottles

Synonym(s):

BGS042

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
976,00 kr.

976,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
976,00 kr.

About This Item

UNSPSC Code:
13111000
NACRES:
NB.43

976,00 kr.


Please contact Customer Service for Availability

feature

to hold up to six 74mm diameter bottles

packaging

pkg of 1 ea

manufacturer/tradename

Azlon BGS042

Looking for similar products? Visit Product Comparison Guide

Application

For safely carrying laboratory bottles

Features and Benefits

Good heat and chemical resistance

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service