Skip to Content
Merck
All Photos(1)

Key Documents

DWK60940A-2

Kimble® 33 Expansion Borosilicate Glass Vial With Attached White Urea Cap And Ptfe-Faced White Rubber Liner

Sign Into View Organizational & Contract Pricing

Select a Size

144 EACH
3.080,00 kr.

3.080,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
144 EACH
3.080,00 kr.

About This Item

UNSPSC Code:
41122001
NACRES:
NB.43

3.080,00 kr.


Please contact Customer Service for Availability

material

Borosilicate glass/PTFE vial
white cap (Urea Cap and PTFE-faced White Rubber Liner)

description

8-425

packaging

pkg of 144 ea

manufacturer/tradename

Kimble® 60940A-2

capacity

2 mL

diam. × H

12 mm × 32 mm

Looking for similar products? Visit Product Comparison Guide

General description

The KIMBLE® 33 Expansion Borosilicate Glass Vial is a clear, screw-thread vial designed for laboratory use, offering both thermal and chemical resistance. It features an attached white urea cap that ensures a secure closure, preventing contamination and evaporation of contents. The vial is lined with a PTFE-faced white rubber liner, enhancing its sealing capabilities and compatibility with various substances. This combination makes it an ideal choice for storing samples, reagents, and other laboratory materials efficiently.

Application

Storage and transport of samples, lyophilization, contain harsh reagents or liquids.

Features and Benefits

  • Closed-top white urea caps with PTFE-faced white rubber liners
  • 60940A-4 complies with EPA 600 series methods
  • Packaged in corrugated trays with partitions
  • Converted from Type I, 33 expansion borosilicate glass tubing.

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are these tubes resistants to -80°C?

    1 answer
    1. This product has not been tested for use at -80°C. However, the item is expected to be suitable. Note that the rubber cap liner hardens at -50°C, the tube must be fully seated prior to reaching this temperature. In addition, the change in temperature, for both the freezing and thawing process, should be incremental and no more than 50° at a time.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service