Skip to Content
Merck
All Photos(4)

Key Documents

DWK292836307

Duran® Gl 45 Bottle Carrying System

Synonym(s):

292836307

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41120000
NACRES:
NB.17

material

red polypropylene

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 292836307

compatibility

use with DURAN® Original GL 45 Bottle

Looking for similar products? Visit Product Comparison Guide

General description

The Bottle Carrying System range fits standard laboratory bottles that have a GL 45 neck finish, and are available for four bottle sizes: 2, 5, 10 or 20 litres. The carriers are ergonomically designed, and offer health and safety benefits to the user. They allow the easy and safe transport of bottles around the laboratory or production facility. All materials are fully autoclavable. The collar that fits to the neck of the bottle is only removable by cutting, so the attachment is 100 % secure in use.

Application

The new modular Bottle Carrying System fi ts standard laboratory bottles that have a GL 45 neck finish and is compatible with four bottle sizes 2, 5, 10 and 20 litres. It is ergonomically designed, and offers health & safety benefits to the user and allows easy and safe transport of bottles around the laboratory or production facility.

Features and Benefits

All materials are fully autoclavable.

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service