Skip to Content
Merck
All Photos(2)

Key Documents

D0628

Sigma-Aldrich

5,6-Dibenzyloxyindole

≥98%

Synonym(s):

5,6-Bis(benzyloxy)indole, NSC 83220

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19NO2
CAS Number:
Molecular Weight:
329.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

storage temp.

−20°C

SMILES string

C(Oc1cc2cc[nH]c2cc1OCc3ccccc3)c4ccccc4

InChI

1S/C22H19NO2/c1-3-7-17(8-4-1)15-24-21-13-19-11-12-23-20(19)14-22(21)25-16-18-9-5-2-6-10-18/h1-14,23H,15-16H2

InChI key

JEUGWNYIRZKHDS-UHFFFAOYSA-N

Application

Reactant for preparation of:
CB2 Cannabinoid Receptor ligands
Potent c-ABL inhibitors
A potent antioxidant agent occuring naturally in beetroot

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service