Key Documents
CLS70360125
Pyrex® VISTA filter flask
capacity 125 mL
Synonym(s):
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 70360
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Product CLS70360125 is not currently sold in your country. Contact Technical Service
material
borosilicate glass
flat bottom flask
narrow-mouth
sterility
non-sterile
feature
closure type ground joint or rubber stopper
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 70360-125
capacity
125 mL
General description
These small filtering flasks are recommended for microchemical use. All flasks have permanent blue enamel marking spots.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)