Skip to Content
Merck
All Photos(2)

Key Documents

C9004

Sigma-Aldrich

Z-DL-Ser-OH

99%

Synonym(s):

N-Cbz-DL-serine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NHCO2CH2C6H5)CO2H
CAS Number:
Molecular Weight:
239.22
Beilstein:
3593625
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

124-126 °C (lit.)

application(s)

peptide synthesis

SMILES string

OCC(NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C11H13NO5/c13-6-9(10(14)15)12-11(16)17-7-8-4-2-1-3-5-8/h1-5,9,13H,6-7H2,(H,12,16)(H,14,15)

InChI key

GNIDSOFZAKMQAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Sypniewski et al.
The Journal of organic chemistry, 65(20), 6595-6600 (2000-10-29)
The enantiospecific synthesis of (R)-Boc-(Fmoc)-aminoglycine 7 was achieved. (S)-Cbz-serine 1 was reacted with diphenylphosphoryl azide in the presence of triethylamine to yield cyclic (S) carbamate 2. The ring nitrogen of 2 was protected with a Boc group (3). The cyclic
Jason Lejeune et al.
Biosensors & bioelectronics, 25(3), 604-608 (2009-03-27)
Changes detected in the imprinting effect by OMNiMIPs imprinted with multiple templates appear to be a function of the maximum template loading. Below the maximum template loading, the polymers imprinted with multiple compounds provide molecular recognition close to the polymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service