Skip to Content
Merck
All Photos(1)

Key Documents

C41703

Sigma-Aldrich

2-Chloroethyl methanesulfonate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3CH2CH2Cl
CAS Number:
Molecular Weight:
158.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

125-126 °C/9 mmHg (lit.)

density

1.39 g/mL at 25 °C (lit.)

SMILES string

CS(=O)(=O)OCCCl

InChI

1S/C3H7ClO3S/c1-8(5,6)7-3-2-4/h2-3H2,1H3

InChI key

FMMYTRQXHORTCU-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N G Fossett et al.
Mutation research, 331(2), 213-224 (1995-10-01)
The 2-chloroethyl methanesulfonate (2ClEMS)-induced alcohol dehydrogenase (Adh) null germline mutation frequency in treated Drosophila melanogaster second instar larval gonia was two orders of magnitude greater than the spontaneous mutation frequency. DNA sequence analysis of 83 Adh null mutations showed that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service