Documents
BR51467
BRAND® ground socket
PTFE, joint: 29/32, grip collar and sealing rings
Synonym(s):
Ground sockets, PTFE
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
material
PTFE
Joint
joint: 29/32
feature
grip collar and sealing rings
manufacturer/tradename
BRAND 51467
Related Categories
General description
PTFE. Greaseless seal will not contaminate sample. Outstanding chemical resistance at operating temperatures from -200 to +260 °C. Sturdy design for permanent use. With grip collar and sealing rings for excellent tightness. Low leakage rate: Q < 1 · 10-4 mbar · l · s-1.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG