Key Documents
BAH195620000
SP Bel-Art® Straight Tubing Connectors
tubing I.D. 0.5-0.5 in. (6-12.7 mm)
Synonym(s):
connectors, tubing, tubing coupler
Select a Size
12 EACH
446,00 kr.
446,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
12 EACH
446,00 kr.
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
446,00 kr.
Please contact Customer Service for Availability
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art H19562-0000
L × width
1.75 in. (44.5 mm) × 0.5625 in. (14.3 mm)
tubing I.D.
0.5-0.5 in. (6-12.7 mm)
General description
These straight fittings ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.).
Features and Benefits
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases.
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)