Documents
BAH131760612
SP Bel-Art® utility bags
width × H × thickness 6 in. (152 mm) × 12 in. (305 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Synonym(s):
clear bag, reusable bag, sample bag
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
clear
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art H13176-0612
width × H × thickness
6 in. (152 mm) × 12 in. (305 mm) × 1.5 mil (0.038 mm)
General description
Reusable, transparent bags are ideal for shipping or storage and are especially useful as sample bags.
Features and Benefits
  • FDA grade polyethylene, safe for food, drug and cosmetic use
  • Latex free
  • Not Autoclavable
  • H13176 lightweight bags are 0.038 mm (1.5 mil) thick, H13178 heavyweight bags are 0.1016 mm (4 mil) thick
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP