BAF420270000
SP Bel-Art® "Space Saver" Polycarbonate Vacuum Desiccator with Clear Polycarbonate Bottom
volume 0.31 ft3
Synonym(s):
SP Bel-Art® "Space Saver" Polycarbonate Vacuum Desiccator with Clear Polycarbonate Bottom
Select a Size
1.600,00 kr.
Select a Size
About This Item
1.600,00 kr.
Recommended Products
material
clear polycarbonate
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F42027-0000
volume
0.31 ft3
General description
- Lighter and less costly than similarly sized glass models; does not chip like glass Holds a full vacuum (29.9" Hg) at room temperature for 24 hours
- Tapered polypropylene stopcock with PTFE plug accepts tubing with ¼ to ⅜” I.D. and allows a controlled entrance of air preventing unnecessary turbulence
- Neoprene O-ring assures a tight seal down to 0˚C without the need for grease (ring will not be harmed if grease is used)
- ‘Flat Dome,’ clear polycarbonate top with large ribbed knob maximizes interior clearances and provides an average of 13% greater interior volume over standard hemispherical domes
- All models include a 0.32 cm (⅛") thick perforated plate; disposable or reusable desiccant can be placed on or under this plate
- White polypropylene bottom models can support incandescent crucibles on Minerit HD High Heat Ceramic Desiccator Plates
- Clear polycarbonate bottom models offer increased viewing visibility from more angles
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)