Key Documents
BAF169590000
SP Bel-Art® Truncated Style Acid/Solvent Bottle Carrier
to hold, one 2.5 Liter(5 pints) Bottle
Synonym(s):
solvent bottle carrier
Select a Size
1 EACH
1.080,00 kr.
1.080,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
1.080,00 kr.
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
1.080,00 kr.
Please contact Customer Service for Availability
feature
to hold one 2.5 Liter(5 pints) Bottle
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F16959-0000
H × diam.
12.5 in. (250 cm) × 7.125 in. (180 cm)
General description
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles.
Features and Benefits
  • Cone style lid has a 16mm (5/8”) opening in the top for insertion of tubing; simplifies the transfer of liquids without pouring
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard US reagent bottles
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)