Key Documents
BAB617005300
SP Bel-Art® Durac® Digital Plastic Stopwatch 1/100 Second Chronograph with Clock Alarm and Calendar
with certificate of calibration
Synonym(s):
Digital Timer, H-B DURAC DIGITAL STOPWATCHES
Select a Size
1 EACH
399,00 kr.
399,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
399,00 kr.
About This Item
UNSPSC Code:
41112303
NACRES:
NB.79
399,00 kr.
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B61700-5300
General description
Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) instructions, electronic stopwatch include a calendar and a clock.
Features and Benefits
  • Features Certificate of Calibration and individual serial number
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST B61700-5200 stores 30 lap times and calculates lap time average, allows for 99 split lap times, and has a 10/320SEC pacer
  • Store 1 lap time and includes an alarm
  • Requires one CR2032 battery (included)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)