Skip to Content
Merck
All Photos(1)

Key Documents

B90807

Sigma-Aldrich

Butyl carbamate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCO2(CH2)3CH3
CAS Number:
Molecular Weight:
117.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

53-55 °C (lit.)

SMILES string

CCCCOC(N)=O

InChI

1S/C5H11NO2/c1-2-3-4-8-5(6)7/h2-4H2,1H3,(H2,6,7)

InChI key

SKKTUOZKZKCGTB-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andrea Y Garzón-Serrano et al.
Journal of forensic sciences, 65(4), 1085-1093 (2020-03-17)
Ammonium nitrate fuel oil is an explosive mixture found in most antipersonnel landmines (APL) buried throughout the Colombian territory. During more than 50 years of internal conflict, the Colombian government has found that trained dogs are the most effective method to
Yong Gun Kim et al.
Food chemistry, 189, 74-79 (2015-07-21)
The aim of this study was to determine the effect of urease on the formation of ethyl carbamate (EC) in the presence of previously known precursors of EC (citrulline, urea, and ethanol) using a soybean paste model system. The levels

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service