Skip to Content
Merck
All Photos(1)

Key Documents

B69204

Sigma-Aldrich

2-Bromoisobutyrophenone

98%

Synonym(s):

2-Bromo-2-methylpropiophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC(CH3)2Br
CAS Number:
Molecular Weight:
227.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5561 (lit.)

bp

146-148 °C/30 mmHg (lit.)

density

1.35 g/mL at 25 °C (lit.)

SMILES string

CC(C)(Br)C(=O)c1ccccc1

InChI

1S/C10H11BrO/c1-10(2,11)9(12)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

QMOSZSHTSOWPRX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin D Smith et al.
Nature communications, 10(1), 1837-1837 (2019-04-25)
Photocatalytic polymers offer an alternative to prevailing organometallics and nanomaterials, and they may benefit from polymer-mediated catalytic and material enhancements. MPC-1, a polymer photoredox catalyst reported herein, exhibits enhanced catalytic activity arising from charge transfer states (CTSs) between its two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service