Skip to Content
Merck
All Photos(1)

Key Documents

ALD00120

Sigma-Aldrich

4′-Methyl-[1,1′-biphenyl]-4-sulfonyl fluoride

95% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11FO2S
CAS Number:
Molecular Weight:
250.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95% (GC)

form

solid

reaction suitability

reaction type: click chemistry

mp

132-137 °C

SMILES string

CC(C=C1)=CC=C1C2=CC=C(S(F)(=O)=O)C=C2

InChI

1S/C13H11FO2S/c1-10-2-4-11(5-3-10)12-6-8-13(9-7-12)17(14,15)16/h2-9H,1H3

InChI key

SRUDIGLPGZXHEO-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service