Skip to Content
Merck
All Photos(1)

Documents

933406

Sigma-Aldrich

Tweezers Protection Box

Synonym(s):

Tweezer case

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

Tweezers Protection Box

Application

Made from soft silicone; grooved properly to tightly hold all types of tweezers in place. The tip of the tweezer is always well protected from any damage. The silicone pad is held in a plastic box with a hinged lid that keeps the tweezers free from dust. Holds 7 tweezers.
Overall measurements: 6⅛"(L) x 4⅛"(W) x 1⅜"(H)
Inside measurements: 6"(L) x 4"(W) x 1¼"(H)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service