Skip to Content
Merck
All Photos(2)

Key Documents

901126

Sigma-Aldrich

trans-Bis(tribenzylphosphine)(2-methylphenyl)nickel(II) chloride

95%

Synonym(s):

trans-(PBn3)2Ni(o-tolyl)Cl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H49ClNiP2
CAS Number:
Molecular Weight:
794.01
UNSPSC Code:
12352200

Assay

95%

form

powder or solid

reaction suitability

reaction type: Cross Couplings
reagent type: catalyst

mp

200-205 °C

SMILES string

Cl[Ni](C1=C(C)C=CC=C1)(P(CC2=CC=CC=C2)(CC3=CC=CC=C3)CC4=CC=CC=C4)P(CC5=CC=CC=C5)(CC6=CC=CC=C6)CC7=CC=CC=C7

Application

The following Ni-complex is an air-stable precatalyst for various Ni-catalyzed reactions which normally require Ni(cod)2, however, this precatalyst will not require any glovebox or schlenk techniques that must be followed with Ni(cod)2. Rates are enhanced and selectivity is maintained when compared to the same reaction with Ni(cod)2.

Legal Information

Patent application PCT/US2014/064565. Sold under license from the Massachusetts Institute of Technology.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in homogeneous nickel catalysis.
Tasker S Z, et al.
Nature, 509(7500), 299-299 (2014)
Sarah Z Tasker et al.
Nature, 509(7500), 299-309 (2014-05-16)
Tremendous advances have been made in nickel catalysis over the past decade. Several key properties of nickel, such as facile oxidative addition and ready access to multiple oxidation states, have allowed the development of a broad range of innovative reactions.

Articles

The Jamison group has developed a library of bench-stable phosphine-containing nickel(II) precatalysts that are converted into active catalysts in situ.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Related Content

Jamison group focuses on new reaction development for organic synthesis, pioneering air-stable nickel precatalysts.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service