Skip to Content
Merck
All Photos(1)

Key Documents

900763

Sigma-Aldrich

2,2,4,4-Tetramethyl-3,8,11,14,17-pentaoxa-2,4-disilaoctadecane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H36O5Si2
CAS Number:
Molecular Weight:
352.61
MDL number:
UNSPSC Code:
26111700

Assay

99%

form

liquid

density

0.929 g/cm3

application(s)

battery manufacturing

Application

Organosilicon electrolytes offer low flammability and high electrochemical and thermal stability compared to the commonly used carbonate based electrolytes, which renders them suitable for lithium ion battery applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased Stability Toward Oxygen Reduction Products for Lithium-Air Batteries with Oligoether-Functionalized Silane Electrolytes.
Zhang Z, et al.
The Journal of Physical Chemistry C, 115, 25535-25542 (2011)
Thermal and electrochemical stability of organosilicon electrolytes for lithium-ion batteries.
Chen X, et al.
Journal of Power Sources, 241, 311-319 (2013)

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service