Skip to Content
Merck
All Photos(1)

Documents

900191

Sigma-Aldrich

Silver Nanoparticle Pneumatic Aerosol® Jet Ink

Synonym(s):

Ag ink, Conductive ink, Silver ink, Xerox® silver nanoparticle pneumatic aerosol® Jet ink, xcm-nsPA-RS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

description

Conductivity: ≥1 x 105 Scm-1
surface tension 28 - 30 mN/m at 25°C

form

dispersion

viscosity

10-13 cP (shear viscosity at 25°C, 1-400 s-1)

cation traces

Ag: 63-67 wt. % (by ash)

storage temp.

2-8°C

Other Notes

Caution, do not freeze.

Legal Information

Aerosol is a registered trademark of Cytec Technology Corp.
Xerox is a registered trademark of Xerox Corporation

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service