Skip to Content
Merck
All Photos(1)

Key Documents

859575

Sigma-Aldrich

S-Boc-2-mercapto-4,6-dimethylpyrimidine

98%

Synonym(s):

O-tert-Butyl-S-(4,6-dimethyl-2-pyrimidinyl) thiocarbonate, Boc-S

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
893,00 kr.

893,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
893,00 kr.

About This Item

Empirical Formula (Hill Notation):
C11H16N2O2S
CAS Number:
Molecular Weight:
240.32
Beilstein:
396984
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

893,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

crystals

mp

48-51 °C (lit.)

functional group

thioether

storage temp.

2-8°C

SMILES string

Cc1cc(C)nc(SC(=O)OC(C)(C)C)n1

InChI

1S/C11H16N2O2S/c1-7-6-8(2)13-9(12-7)16-10(14)15-11(3,4)5/h6H,1-5H3

InChI key

POTDIELOEHTPJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service