Skip to Content
Merck
All Photos(1)

Key Documents

855006

Sigma-Aldrich

Thymidine

≥99%

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 2′-Deoxythymidine, Thymine deoxyriboside, dT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
Beilstein:
89285
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

≥99%

optical activity

[α]25/D +18.5°, c = 1 in H2O

mp

186-188 °C (lit.)

SMILES string

CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m0/s1

InChI key

IQFYYKKMVGJFEH-XLPZGREQSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yee Suan Poo et al.
PLoS neglected tropical diseases, 8(12), e3354-e3354 (2014-12-05)
The recent epidemic of the arthritogenic alphavirus, chikungunya virus (CHIKV) has prompted a quest to understand the correlates of protection against virus and disease in order to inform development of new interventions. Herein we highlight the propensity of CHIKV infections
Anne P Bouvy et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 29(8), 1587-1597 (2014-04-29)
To evaluate the influence of induction therapy on Tregs we investigated their origin, kinetics and function in kidney transplant patients after treatment with T-cell depleting rabbit antithymocyte globulin (rATG) or IL-2 receptor antagonist basiliximab. Flow cytometry was used to study
Johan Vande Voorde et al.
The Journal of biological chemistry, 289(19), 13054-13065 (2014-03-29)
The intracellular metabolism and cytostatic activity of the anticancer drug gemcitabine (2',2'-difluoro-2'-deoxycytidine; dFdC) was severely compromised in Mycoplasma hyorhinis-infected tumor cell cultures. Pronounced deamination of dFdC to its less cytostatic metabolite 2',2'-difluoro-2'-deoxyuridine was observed, both in cell extracts and spent
Athan C Baillet et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(6), 1535-1547 (2015-01-28)
Chlamydia trachomatis is a sexually transmitted obligate intracellular pathogen that causes inflammatory reactive arthritis, spondylitis, psoriasiform dermatitis, and conjunctivitis in some individuals after genital infection. The immunologic basis for this inflammatory response in susceptible hosts is poorly understood. As ZAP-70(W163C)
Paul Mésange et al.
Oncotarget, 5(13), 4709-4721 (2014-07-13)
Colorectal cancer (CRC) is a common tumor type with a high mortality rate, in part due to intrinsic drug resistance. Although bevacizumab, a VEGF-directed neutralizing antibody, is particularly active in this pathology, some patients never respond for reasons not well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service