Skip to Content
Merck
All Photos(1)

Documents

805769

Sigma-Aldrich

1-Butene-d8

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2=CDCD2CD3
Molecular Weight:
64.16
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

gas

mass shift

M+8

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])=C([2H])[2H]

InChI

1S/C4H8/c1-3-4-2/h3H,1,4H2,2H3/i1D2,2D3,3D,4D2

InChI key

VXNZUUAINFGPBY-BGWBEDHISA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-112.0 Ā°F - closed cup

Flash Point(C)

-80 Ā°C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service