Skip to Content
Merck
All Photos(3)

Key Documents

803774

Sigma-Aldrich

Colby trifluoromethylation reagent

Synonym(s):

Hexafluoroacetone hydrate-1,8-diazo-bicyclo[5.4.0]undec-7-ene salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18F6N2O2
CAS Number:
Molecular Weight:
336.27
UNSPSC Code:
12352101

form

solid

SMILES string

OC([O-])(C(F)(F)F)C(F)(F)F.C1CCC2=[N+](CC1)CCCN2

Looking for similar products? Visit Product Comparison Guide

General description

Colby trifluoromethylation reagent is an amidinate salt of hexafluoroacetone hydrate. This reagent can be prepared from the ethereal solution of hexafluoroacetone trihydrate. It undergoes various trifluoromethylation reactions with electrophiles under different reaction conditions to afford trifluoromethylated products.

Application

Colby trifluoromethylation reagent may be used for the preparation of fluorinated organic compounds.
The Colby Trifluoromethylation Reagent is an air-stable, white solid, which can be used as a source of fluoroform through the release of trifluoroacetate. This reagent can add trifluoromethyl groups in a nucleophlic fashion to carbonyls and sulfides.

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark V Riofski et al.
Organic letters, 15(1), 208-211 (2012-12-18)
A powerful, new reagent, an amidinate salt of hexafluoroacetone hydrate, is an air-stable salt that can be used for the preparation of fluorinated organic molecules. Nucleophilic trifluoromethylation reactions are demonstrated following the base-promoted release of trifluoroacetate. This reagent is soluble

Related Content

Prof. David Colby's laboratory is developing synthetic methods to enable the efficient modification of complex molecules, such as natural products, as well as the production of fluorinated compounds. They have focused on understanding how to cleave bonds in order to remodel structures, and their first major discovery is that the release of trifluoroacetate is a powerful approach to break carbon–carbon bonds. This strategy has enabled the invention of new reagents and methods for the synthesis of fluorinated products for drug discovery and for exploring medicinal chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service