Skip to Content
Merck
All Photos(2)

Key Documents

802441

Sigma-Aldrich

Diphenylphosphine selenide

95%

Synonym(s):

DPPSe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11PSe
CAS Number:
Molecular Weight:
265.15
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder or crystals

grade

for analytical purposes

Assay

95%

form

powder or crystals

mp

110-115 °C

storage temp.

15-25°C

SMILES string

[Se]=P(C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C12H11PSe/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H

InChI key

KDFCMIXBMVVIQJ-UHFFFAOYSA-N

Application

The chalcogenide based quantum dots find application in photovoltaics, light emitting diodes and precursor in synthesis of the chalcogenides based quantum dots.[1][2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandi M Cossairt et al.
The journal of physical chemistry letters, 2(4), 3075-3080 (2012-01-10)
A series of nonstoichiometric CdSe clusters with lowest energy electronic absorptions between 409 - 420 nm has been prepared from cadmium 1-naphthoate, 2-naphthoate, 4-thiomethyl-1-naphthaote, and 1-naphthalene thiolate complexes and diphenylphosphine selenide (DPPSe). Pair distribution function analysis of X-ray diffraction data
Mysteries of TOPSe Revealed: Insights into Quantum Dot Nucleation.
Evans CM, et al.
Journal of the American Chemical Society, 132, 3075-3080 (2010)

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service