Skip to Content
Merck
All Photos(1)

Key Documents

793868

Sigma-Aldrich

1-[(4-Methylphenyl)sulfonyl]-3-azetidinone

95%

Synonym(s):

1-(p-Tosyl)azetidin-3-one, N-Tosyl-3-azetidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO3S
CAS Number:
Molecular Weight:
225.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

147-152 °C

functional group

ketone
sulfonamide

storage temp.

2-8°C

SMILES string

O=C1CN(S(C2=CC=C(C)C=C2)(=O)=O)C1

InChI

1S/C10H11NO3S/c1-8-2-4-10(5-3-8)15(13,14)11-6-9(12)7-11/h2-5H,6-7H2,1H3

InChI key

YRVBXEVVGIQJOZ-UHFFFAOYSA-N

Application

Protected azetidinone has shown to be a versatile building block in the formation of functionalized spirocyclic oxetanes by the Carreira group.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service