Skip to Content
Merck
All Photos(1)

Documents

792659

Sigma-Aldrich

3-Hydroxypropionic acid solution

30% in water

Synonym(s):

3-Hydroxypropanoic acid solution, 3-Hydroxypropanoic acid, Hydracrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product name

3-Hydroxypropionic acid, 30%solution in water,

form

liquid

concentration

30% in water

refractive index

n/D 1.364

density

1.066 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OC(CCO)=O

InChI

1S/C3H6O3/c4-2-1-3(5)6/h4H,1-2H2,(H,5,6)

InChI key

ALRHLSYJTWAHJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Chemical synthesis: used as a precursor for several higher value-added chemical products (Garg et al., 2023).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service