Skip to Content
Merck
All Photos(1)

Documents

777706

Sigma-Aldrich

4-(Difluoromethoxy)-3-hydroxybenzaldehyde

95%

Synonym(s):

3-Hydroxy-4-difluoromethoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6F2O3
CAS Number:
Molecular Weight:
188.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

solid

mp

85-90 °C

SMILES string

FC(F)OC1=C(O)C=C(C([H])=O)C=C1

InChI

1S/C8H6F2O3/c9-8(10)13-7-2-1-5(4-11)3-6(7)12/h1-4,8,12H

InChI key

ZLIKNROJGXXNJG-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service