Skip to Content
Merck
All Photos(1)

Documents

774472

Sigma-Aldrich

Ethyl 2-(4-methoxyphenylcarbonothioylthio)acetate

99%

Synonym(s):

2-[(4-Methoxyphenylthioxomethyl)thio]ethanoic acid ethyl ester, CTA, RAFT dithiobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O3S2
CAS Number:
Molecular Weight:
270.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

mp

30-35 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)CSC(=S)c1ccc(OC)cc1

InChI

1S/C12H14O3S2/c1-3-15-11(13)8-17-12(16)9-4-6-10(14-2)7-5-9/h4-7H,3,8H2,1-2H3

InChI key

YZSCQGUNDFCXBR-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of methacrylate and methacrylamide monomers. Chain Transfer Agent (CTA) with electron donating group to increase fragmentation and reduce polymerization time.

Other Notes

Reversible Addition Fragmentation Chain Transfer (RAFT) Polymerization. Please register for your free Controlled Radical Polymerization Guide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brendan Quiney et al.
Journal of computer assisted tomography, 41(6), 849-853 (2017-05-10)
The aim of this article was to evaluate the association of intracranial artery calcification (IAC) with acute downstream ischemic stroke (dAIS)/transient ischemic attack while considering stenosis. Consecutive stroke computed tomography angiography head/neck examinations from January 2010 to April 2010 were
Volker Maus et al.
Cerebrovascular diseases (Basel, Switzerland), 45(1-2), 10-17 (2017-12-07)
Cerebral ischemic strokes due to extra-/intracranial tandem occlusions (TO) of the anterior circulation are responsible for causing mechanical thrombectomy (MT). The impact of concomitant contralateral carotid stenosis (CCS) upon outcome remains unclear in this stroke subtype. Retrospective analysis of prospectively
Trisha L Roy et al.
Journal of vascular surgery, 67(6), 1844-1854 (2017-12-19)
Limitations with current peripheral arterial imaging modalities make selection of patients for percutaneous vascular interventions difficult. The purpose of this study was to determine whether a novel preprocedural magnetic resonance imaging (MRI) method can identify lesions that would be more
Aukelien C Dimitriu-Leen et al.
JACC. Cardiovascular imaging, 10(12), 1451-1458 (2017-05-22)
The aim of the present study was to evaluate, in low-to-intermediate pre-test probability patients who were referred for coronary computed tomography angiography (CTA) and did not show obstructive coronary artery disease (CAD), whether an intramural course of a coronary artery
Francesco Scardulla et al.
The Journal of heart and lung transplantation : the official publication of the International Society for Heart Transplantation, 36(8), 906-913 (2017-04-23)
The use of left ventricular assist devices (LVADs) to treat advanced cardiac heart failure is constantly increasing, although this device leads to high risk for gastrointestinal bleeding. Using in-silico flow analysis, we quantified hemodynamic alterations due to continuous-flow LVAD (HeartWare

Articles

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service