Skip to Content
Merck
All Photos(2)

Key Documents

771600

Sigma-Aldrich

Zirconium(IV) dibutoxide(bis-2,4-pentanedionate) solution

25 wt. % (in 1-butanol/toluene), 99.9% trace metals basis

Synonym(s):

Bis(acetylacetonato)dibutoxyzirconium, Dibutoxybis(acetylacetonato)zirconium, Dibutoxyzirconium bis(acetylacetonate), Zirconium bis(n-butoxy) bis(acetoacetonate), Zirconium di-n-butoxide(bis-2,4-pentanedionate), Zirconium, dibutoxybis(2,4-pentanedionato), Zirconiumdibutoxide bis(2,4-pentanedionate), Zirconiumdibutoxybisacetylacetonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O6Zr
Molecular Weight:
435.67
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99.9% trace metals basis

form

liquid

reaction suitability

core: zirconium

concentration

25 wt. % (in 1-butanol/toluene)

refractive index

n20/D 1.487

density

0.928 g/mL at 25 °C

SMILES string

CCCCO[Zr](OCCCC)(O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.2C4H9O.Zr/c2*1-4(6)3-5(2)7;2*1-2-3-4-5;/h2*3,6H,1-2H3;2*2-4H2,1H3;/q;;2*-1;+4/p-2/b2*4-3-;;;

InChI key

VKWRHWOICYRNLR-VGKOASNMSA-L

Application

Zirconium di-n-butoxide(bis-2; 4-pentanedionate) is a metal organic precursor of zirconium suitable for the preparation of nanoparticles; coatings and thin films containing ZrO2.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F

Flash Point(C)

8 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The precise synthesis and growth of core-shell nanoparticles within a self-assembled spherical template.
Kosuke Suzuki et al.
Angewandte Chemie (International ed. in English), 50(21), 4858-4861 (2011-03-05)

Articles

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service