Skip to Content
Merck
All Photos(2)

Documents

767360

Sigma-Aldrich

5-Ethynyl-1,3-benzenedicarboxylic acid

greener alternative

99% (HPLC)

Synonym(s):

5-Ethynylisophthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6O4
CAS Number:
Molecular Weight:
190.15
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (HPLC)

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

245-255 °C

greener alternative category

SMILES string

OC(=O)c1cc(cc(c1)C(O)=O)C#C

InChI

1S/C10H6O4/c1-2-6-3-7(9(11)12)5-8(4-6)10(13)14/h1,3-5H,(H,11,12)(H,13,14)

InChI key

XKEUZQRIANAGPB-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

MOF linker used for the synthesis of MOFs with pores which can be further functionalized by click chemistry

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface functionalization of porous coordination nanocages via click chemistry and their application in drug delivery.
Dan Zhao et al.
Advanced materials (Deerfield Beach, Fla.), 23(1), 90-93 (2010-10-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service