Skip to Content
Merck
All Photos(1)

Key Documents

767263

Sigma-Aldrich

Polyester bis-MPA dendron, 8 hydroxyl, 1 amine

generation 3

Synonym(s):

bisMPA dendrimer, bisMPA dendron

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

hydroxyl surface groups

form

solid

mol wt

generation 3

feature

focal point amine

no. Surface Groups

8

mp

275-280 °C

storage temp.

−20°C

SMILES string

CCC(COC(=O)c1ccc(N)cc1)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO

InChI

1S/C43H67NO22/c1-8-43(25-60-30(53)28-9-11-29(44)12-10-28,26-65-35(58)41(6,21-61-31(54)37(2,13-45)14-46)22-62-32(55)38(3,15-47)16-48)27-66-36(59)42(7,23-63-33(56)39(4,17-49)18-50)24-64-34(57)40(5,19-51)20-52/h9-12,45-52H,8,13-27,44H2,1-7H3

InChI key

ABFRDJWLXWYYKC-UHFFFAOYSA-N

General description

calculated mol. wt. 950

Application

Amine dendrons can be covalently attached through conjugation to carboxyl groups, primary amines, or sulfhydryl groups. Amines can also attach to oxidized carbohydrate groups (usually aldehydes) by reductive amination.

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service