Skip to Content
Merck
All Photos(1)

Key Documents

759791

Sigma-Aldrich

2,5-Dibromo-3,4-ethylenedioxythiophene

97%

Synonym(s):

5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Br2O2S
CAS Number:
Molecular Weight:
299.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

95-100 °C

storage temp.

−20°C

SMILES string

Brc1sc(Br)c2OCCOc12

InChI

1S/C6H4Br2O2S/c7-5-3-4(6(8)11-5)10-2-1-9-3/h1-2H2

InChI key

FHMRWRBNAIDRAP-UHFFFAOYSA-N

General description

2,5-Dibromo-3,4-ethylenedioxythiophene (DBEDOT) is a monomer which is used in the synthesis of highly conducting poly(3,4-ethylenedioxythiophene) (PEDOT) polymer. This polymer is formed by solid-state polymerisation of DBEDOT and is used widely in organic-light emitting diodes and polymer field effect transistors. DBEDOT is a well-ordered crystalline monomer and produces PEDOT polymer which has a high degree of order. [1]

Application

Used as a monomer in the synthesis of poly(3,4-ethylenedioxythiophene) (PEDOT) polymer which are used in electrochromic devices.[2]

Features and Benefits

Polymerisation of DBEDOT can take place under very gentle conditions in the absence of external agents or catalysts.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of thiophene functionalized polystyrene copolymers and their electrochemical properties
Sahin, E., Camurlu, P., Toppare, L., Mercore, V. M., Cianga, I., & Yagci, Y.
Polymer International, 54(12), 1599-1605 (2005)
Hong Meng et al.
Journal of the American Chemical Society, 125(49), 15151-15162 (2003-12-05)
Prolonged storage ( approximately 2 years) or gentle heating (50-80 degrees C) of crystalline 2,5-dibromo-3,4-ethylenedioxythiophene (DBEDOT) affords a highly conducting, bromine-doped poly(3,4-ethylenedioxythiophene) (PEDOT), as confirmed by solid-state NMR, FTIR, CV, and vis-NIR spectroscopies. The novel solid-state polymerization (SSP) does not
Minami Kato et al.
ChemSusChem, 13(9), 2379-2385 (2020-02-11)
Many types of batteries have been proposed as next-generation energy-storage systems. One candidate is a rocking-chair-type "molecular ion battery" in which a molecular ion, instead of Li+ , works as a charge carrier. Previously, we reported a viologen-type derivative as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service