Skip to Content
Merck
All Photos(1)

Key Documents

752614

Sigma-Aldrich

Silicon aluminum oxynitride

99.9% trace metals basis

Synonym(s):

Aluminum silicon nitride oxide, Sialon, Silica aluminum nitride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al6N6O2Si
CAS Number:
Molecular Weight:
306.01
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.9% trace metals basis

form

powder

mp

1800 °C (lit.)

SMILES string

O1[Al]2N=[Al]N3[Al]=N[Al]4O[Si]13N([Al]=N2)[Al]=N4

InChI

1S/6Al.N2O2Si.4N/c;;;;;;1-5(2,3)4;;;;/q;;;;2*+1;-2;;;;

InChI key

UXCKNSFMFVWLRV-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen, I. W.; Rosenflanz, A.
Nature, 389, 701-701 (1997)
Ekstrom, T.; Nygren, M.
Journal of the American Ceramic Society. American Ceramic Society, 75, 259-259 (1992)
Bandyopadhyay, S.
J. Eur. Ceram. Soc., 17, 929-929 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service