Skip to Content
Merck
All Photos(1)

Key Documents

750581

Sigma-Aldrich

Thieno[3,2-b]thiophene-2-carboxaldehyde

98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4OS2
CAS Number:
Molecular Weight:
168.24
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98% (HPLC)

form

powder or crystals

mp

52-56 °C

SMILES string

O=Cc1cc2sccc2s1

InChI

1S/C7H4OS2/c8-4-5-3-7-6(10-5)1-2-9-7/h1-4H

InChI key

JKVPCDAKZCFJBU-UHFFFAOYSA-N

General description

This molecule has been used in the synthesis of novel metal-free organic dyes for Dye-Sensitized Solar Cells reaching conversion efficiencies of 6.23%.†

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-Lu Li et al.
Chemical communications (Cambridge, England), 26(26), 2792-2794 (2006-10-03)
Two novel metal-free organic dyes containing thienothiophene and thiophene segments have been synthesized. Nano-crystalline TiO2 dye-sensitized solar cells were fabricated using these dyes as light-harvesting sensitizers, and a high solar energy-to-electricity conversion efficiency of 6.23% was achieved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service