744247
(1R,2S)-cis-N-Boc-2-aminocyclohexanol
≥97% (GC)
Synonym(s):
(1R,2S)-N-tert-Butoxycarbonyl-2-aminocyclohexanol, tert-Butyl (1S,2R)-2-hydroxycyclohexylcarbamate
About This Item
Assay
≥97% (GC)
form
powder
optical purity
enantiomeric excess: ≥97%
SMILES string
CC(C)(C)OC(=O)N[C@H]1CCCC[C@H]1O
InChI
1S/C11H21NO3/c1-11(2,3)15-10(14)12-8-6-4-5-7-9(8)13/h8-9,13H,4-7H2,1-3H3,(H,12,14)/t8-,9+/m0/s1
InChI key
XVROWZPERFUOCE-DTWKUNHWSA-N
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service