Skip to Content
Merck
All Photos(1)

Key Documents

743585

Sigma-Aldrich

(S)-(−)-1-(2-Diphenylphosphino-1-naphthyl)isoquinoline

≥95.0% (qNMR)

Synonym(s):

(S)-QUINAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H22NP
CAS Number:
Molecular Weight:
439.49
Beilstein:
6664856
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (qNMR)

form

crystals

optical purity

enantiomeric excess: ≥98.0%

storage temp.

2-8°C

InChI

1S/C31H22NP/c1-3-13-25(14-4-1)33(26-15-5-2-6-16-26)29-20-19-23-11-7-9-17-27(23)30(29)31-28-18-10-8-12-24(28)21-22-32-31/h1-22H

InChI key

YMJAIEYASUCCMJ-UHFFFAOYSA-N

General description

(S)-QUINAP is a bidentate ligand used in catalytic asymmetric hydroboration, hydrogenation, oxidation, and allylic alkylation reactions.

Application

(S)-QUINAP can be used as a ligand:
  • In the asymmetric hydrosilylation of alkyl and α, β-unsaturated ketones in the presence of Fe(OAc)2.      
  • For the synthesis of chiral organoboron compounds by copper-catalyzed β-borylation of α,β-unsaturated esters.

[Ir(cod)Cl]2 treated with ligand (S)-QUINAP forms an iridium complex. This complex is used as a catalyst for the enantioselective hydrogenation of olefins. (S)-QUINAP–Rh complex can be employed as a catalyst in enantioselective hydroboration/oxidation of perfluoroalkenes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William J Fleming et al.
Organic & biomolecular chemistry, 7(12), 2520-2524 (2009-06-09)
The synthesis and resolution of a new axially chiral Quinazolinap ligand are reported. The application of this and other related P-N ligands to the copper catalyzed beta-borylation of alpha,beta-unsaturated esters resulted in conversions of up to 100% and ee values
Enantioselective hydrogenation of olefins with axial chiral iridium QUINAP complex
Li X, et al.
Tetrahedron Letters, 48(22), 3915-3917 (2007)
Anna M Segarra et al.
Chemical communications (Cambridge, England), (4), 464-465 (2004-02-07)
Enantioselective access to Markovnikov regioisomeric perfluoroalcohols is achieved in the presence of chiral cationic rhodium complexes and specific hydroborating reagents.
Practical Preparation and Resolution of 1-(2 `-Diphenylphosphino-1 `-naphthyl) isoquinoline: A Useful Ligand for Catalytic Asymmetric Synthesis
Lim CW, et al.
Organic Process Research & Development, 7(3), 379-384 (2003)
Iron-catalyzed enantioselective hydrosilylation of ketones.
Nadim S Shaikh et al.
Angewandte Chemie (International ed. in English), 47(13), 2497-2501 (2008-02-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service