Skip to Content
Merck
All Photos(1)

Documents

743259

Sigma-Aldrich

1,3-Bis[(1S)-2,2-dimethyl-1-(1-naphthyl)propyl]imidazolinium iodide

≥96% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H39IN2
CAS Number:
Molecular Weight:
590.58
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥96% (HPLC)

form

powder

reaction suitability

reagent type: catalyst

impurities

≤1.0% water

SMILES string

[I-].CC(C)(C)[C@H](N1CC[N+](=C1)[C@@H](c2cccc3ccccc23)C(C)(C)C)c4cccc5ccccc45

InChI

1S/C33H39N2.HI/c1-32(2,3)30(28-19-11-15-24-13-7-9-17-26(24)28)34-21-22-35(23-34)31(33(4,5)6)29-20-12-16-25-14-8-10-18-27(25)29;/h7-20,23,30-31H,21-22H2,1-6H3;1H/q+1;/p-1/t30-,31+;

InChI key

FBAXFNUYKXNWGQ-USHPRDJASA-M

Application

Carbene ligand precursor

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fused indolines by palladium-catalyzed asymmetric C-C coupling involving an unactivated methylene group.
Masafumi Nakanishi et al.
Angewandte Chemie (International ed. in English), 50(32), 7438-7441 (2011-07-05)
Nakanishi, M.; et al.
Angewandte Chemie (International Edition in English), 50, 9775-9775 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service