Skip to Content
Merck
All Photos(1)

Key Documents

742457

Sigma-Aldrich

1-(Azidomethyl)-4-bromobenzene solution

~0.5 M in dichloromethane, ≥96.0% (HPLC)

Synonym(s):

p-Bromobenzyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6BrN3
CAS Number:
Molecular Weight:
212.05
Beilstein:
4904917
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥96.0% (HPLC)

form

solution

reaction suitability

reaction type: click chemistry

concentration

~0.5 M in dichloromethane

impurities

≤2.0% water

storage temp.

2-8°C

SMILES string

Brc1ccc(CN=[N+]=[N-])cc1

InChI

1S/C7H6BrN3/c8-7-3-1-6(2-4-7)5-10-11-9/h1-4H,5H2

InChI key

MOKKTDQWMSOCAF-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Dermal - STOT SE 3

Target Organs

Central nervous system, Lungs,Nervous system,Cardio-vascular system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew D Graaf et al.
ACS chemical biology, 11(10), 2829-2837 (2016-10-22)
Cu(I)-catalyzed "click" reactions cannot be performed on a borate ester derived polymer coating on a microelectrode array because the Cu(II) precursor for the catalyst triggers background reactions between both acetylene and azide groups with the polymer surface. Fortunately, the Cu(II)-background

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service